Dråbefangere, Stutzer, lige
14/23 NS, 14/23 NS
Katalog # 271-2233
Leverandør: Lenz Laborglas
Gå til:
Product Details & Documents
DURAN®, borosilicate 3.3 glass, clear.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Anbringes direkte på destillationskolben for at fange dråber, med to standardslib.
Documents
BSE TSE Glass Columns
BSE/TSE-certifikater
Specifikationer
- Konusstr.:14/23 NS
- Kappestr.:14/23 NS
- Seq:01