Orders LinkContactUsLinkComponent
Skilletragte, Dråbeformet
Skilletragte, Dråbeformet
1000 ml, 29/32 NS, 20 ml
Catalog # 527-1305
Supplier:   Lenz Laborglas
Skilletragte, Dråbeformet
Catalog # 527-1305
Supplier:  Lenz Laborglas
Skilletragte, Dråbeformet
Skilletragte, Dråbeformet
1000 ml, 29/32 NS, 20 ml
Price
932,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
DURAN®, borosilikatglas, med NS-PE prop.
With socket and plain stem. Available with interchangeable solid glass key or alternatively PTFE key for grease-free performance. With or without graduations.
Documents
Specifications
  • Kappestr.:
    29/32 NS
  • Kapacitet:
    1000 ml
  • Inddeling:
    20 ml