Orders LinkContactUsLinkComponent
Koniske skilletragte
Koniske skilletragte
100 ml, 19/26 NS, 5 ml
Catalog # 527-1202
Supplier:   Lenz Laborglas
Koniske skilletragte
Catalog # 527-1202
Supplier:  Lenz Laborglas
Koniske skilletragte
Koniske skilletragte
100 ml, 19/26 NS, 5 ml
Price
550,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
DURAN®, borosilikatglas, med NS-PE prop.
With socket and plain stem. Available with interchangeable solid glass key or alternatively PTFE key for grease-free performance. Both with or without graduations.
Documents
Specifications
  • Kappestr.:
    19/26 NS
  • Kapacitet:
    100 ml
  • Inddeling:
    5 ml