Fork clamps for spherical joints
35
Catalog # 201-7344
Supplier: Lenz Laborglas
Gå til:
Product Details & Documents
Clips for spherical joints made from stainless steel chrome/nickel.
- Easy to install and remove
- Excellent heat and chemical resistance
- With set screw
Specifikationer
- Til NS:35
- Seq:02