Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Spritlampe, DURAN®

Stik, 75×103 mm
  451-3112
Spritlampe, DURAN®
  451-3112
Spritlampe, DURAN®
Spritlampe, DURAN®

Stik, 75×103 mm

 
14,10 kr
   
1 * 1 Items
 

Soda-lime glass with ground glass joint cap. The spirit lamp is an alternative to the gas-powered Bunsen burner and is also suitable for heating cooking glasses and the materials they contain.

Kapacitet: 100 ml

Ø×H: 75×103 mm.

 : Leveres uden sokkel og væge.

 
  • Beskrivelse:
    Stik
  • Seq:
    03
  • Diameter×H:
    75×103 mm