Adapters, expansion, DURAN®
Expansion, 19/26 NS, 29/32 NS
SCOT286548107
DURAN®, borosilicate glass 3,3 clear.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Expansion adapter from small cone to large socket.
- Konusstr.:19/26 NS
- Kappestr.:29/32 NS
- Model:Expansion
- Seq:05