Orders LinkContactUsLinkComponent
Propper, kort slib
24/20
  217-0805
Propper, kort slib
  217-0805
Propper, kort slib
Propper, kort slib
24/20
3.400,00 kr
1 * 100 Items
DURAN®, borosilicate glass 3.3, clear.
  • Hexagonal
  • Short ground
  • NS:
    24/20