Propper, kort slib
12/14
Catalog # 217-0799
Supplier: DWK Life Sciences
Jump to:
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear.
- Hexagonal
- Short ground
Specifications
- NS:12/14