Orders LinkContactUsLinkComponent
Propper, diamantslebne
Propper, diamantslebne
7/16
Katalog # 217-0796
Leverandør:   DWK Life Sciences
Propper, diamantslebne
Katalog # 217-0796
Leverandør:  DWK Life Sciences
Propper, diamantslebne
Propper, diamantslebne
7/16
Price
2.680,00 kr
Log ind for tilgængelighed
1 * 100 Items
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear.
  • Hexagonal
  • Flat
  • Diamond-ground
Specifikationer
  • NS:
    7/16
  • Seq:
    01