Orders LinkContactUsLinkComponent
Ubbelohde viscometers (ASTM)
Ubbelohde viscometers (ASTM)
6000 - 30 000 mm²/s, 285 mm, 4,32 mm
Katalog # SCOI285400801
Leverandør:   SI Analytics
Ubbelohde viscometers (ASTM)
Katalog # SCOI285400801
Leverandør:  SI Analytics
Ubbelohde viscometers (ASTM)
Ubbelohde viscometers (ASTM)
6000 - 30 000 mm²/s, 285 mm, 4,32 mm
Price
3.510,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Ubbelohde viscometers for manual measurements of transparent liquids.
Certifications: ASTM
Specifikationer
  • Område:
    6000 - 30 000 mm²/s
  • Længde:
    285 mm
  • Seq:
    11
  • Ø indv.:
    4,32 mm