Kabler til elektroder
Plug cable combination, A 1 A (DIN instrument plug, 1 m coaxial cable, DIN instrument plug), 1 m
Katalog # 662-2197
Leverandør: SI Analytics
Gå til:
Product Details & Documents
Documents
Specifikationer
- Beskrivelse:Plug cable combination, A 1 A (DIN instrument plug, 1 m coaxial cable, DIN instrument plug)
- Længde:1 m
- Seq:08