Orders LinkContactUsLinkComponent
Adaptere med standardslib, DURAN®
ekspansion, 14/23 NS, 29/32 NS
  SCOT241144208
Adaptere med standardslib, DURAN®
  SCOT241144208
Adaptere med standardslib, DURAN®
Adaptere med standardslib, DURAN®
ekspansion, 14/23 NS, 29/32 NS
272,00 kr
1 * 1 Items
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Adapters, reduction or expansion, with standard ground conical joints.
 : DIN 12257
  • Konusstr.:
    14/23 NS
  • Kappestr.:
    29/32 NS
  • Model:
    ekspansion