Adaptere med standardslib, DURAN®
ekspansion, 14/23 NS, 29/32 NS
SCOT241144208
DURAN®, borosilicate glass 3.3, clear.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Adapters, reduction or expansion, with standard ground conical joints.
: DIN 12257
- Konusstr.:14/23 NS
- Kappestr.:29/32 NS
- Model:ekspansion