Adaptere med standardslib, DURAN®
Ekspansion, 19/26 NS, 24/29 NS
201-1704
DURAN®, borosilicate glass 3.3, clear.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Adapters, reduction or expansion, with standard ground conical joints.
: DIN 12257
- Konusstr.:19/26 NS
- Kappestr.:24/29 NS
- Model:Ekspansion