Orders LinkContactUsLinkComponent
Adaptere med standardslib, DURAN®
Ekspansion, 19/26 NS, 24/29 NS
  201-1704
Adaptere med standardslib, DURAN®
  201-1704
Adaptere med standardslib, DURAN®
Adaptere med standardslib, DURAN®
Ekspansion, 19/26 NS, 24/29 NS
279,00 kr
1 * 1 Items
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Adapters, reduction or expansion, with standard ground conical joints.
 : DIN 12257
  • Konusstr.:
    19/26 NS
  • Kappestr.:
    24/29 NS
  • Model:
    Ekspansion