Orders LinkContactUsLinkComponent
Urglas, DURAN®
Urglas, DURAN®
Watch glass, DURAN® borosilicate glass, with fused edges, 40 mm
Catalog # 216-1110
Supplier:   DWK Life Sciences
Urglas, DURAN®
Catalog # 216-1110
Supplier:  DWK Life Sciences
Urglas,  DURAN®
Urglas, DURAN®
Watch glass, DURAN® borosilicate glass, with fused edges, 40 mm
Price
95,40 kr
Log in for availability
1 * 1 Items
Product Details & Documents
DURAN®, borosilikatglas 3.3, klar, med sammensmeltede kanter.
  • Meget god kemisk resistens
  • Høj temperaturresistens
  • Minimal varmeudvidelse giver relativt høj resistens overfor temperaturændringer
DIN 12341
 : DIN 12341
Specifications
  • Ø:
    40 mm
  • Beskrivelse:
    Watch glass, DURAN® borosilicate glass, with fused edges
  • Seq:
    01