Orders LinkContactUsLinkComponent
DURAN® Blanks for Evaporating Flasks
DURAN® Blanks for Evaporating Flasks
250 ml
Katalog # SCOT210283608
Leverandør:   DWK Life Sciences
DURAN® Blanks for Evaporating Flasks
Katalog # SCOT210283608
Leverandør:  DWK Life Sciences
DURAN® Blanks for Evaporating Flasks
DURAN® Blanks for Evaporating Flasks
250 ml
Price
1.770,00 kr
Log ind for tilgængelighed
1 * 10 Items
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
For glass manipulators without joint.
Specifikationer
  • Kapacitet:
    250 ml