DURAN® Blanks for Evaporating Flasks
100 ml
Katalog # SCOT210282406
Leverandør: DWK Life Sciences
Gå til:
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
For glass manipulators without joint.
Specifikationer
- Kapacitet:100 ml
- Seq:01