Orders LinkContactUsLinkComponent
Propper til separationstragte, DURAN®
Propper til separationstragte, DURAN®
50 ml
Catalog # SCOT210131707
Supplier:   DWK Life Sciences
Propper til separationstragte, DURAN®
Catalog # SCOT210131707
Supplier:  DWK Life Sciences
Propper til separationstragte, DURAN®
Propper til separationstragte, DURAN®
50 ml
Price
1.350,00 kr
Log in for availability
1 * 10 Items
Product Details & Documents
Made of DURAN®, borosilicate glass 3.3, clear.
  • For glass manipulators
  • Very good chemical resistance
  • Very good temperature resistance
Specifications
  • Kapacitet:
    50 ml