Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Centrifugerør

Centrifugerør

15 ml, 17×113 mm
Katalog # 525-1242
Leverandør:   Brand
Centrifugerør
Katalog # 525-1242
Leverandør:  Brand
Centrifugerør
Centrifugerør

15 ml, 17×113 mm

Price
592,00 kr
Log ind for tilgængelighed
1 * 100 Items
Product Details & Documents

AR® glass, conical bottom, beaded rim (except 525-0500 cone-pointed bottom, rimless).

  • Tåler belastninger op til 3000×g
Specifikationer
  • Ø×L:
    17×113 mm
  • Kapacitet:
    15 ml
  • Seq:
    01
Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.