Cavity slides
Cavity slides, klar, 1, 15 - 18 mm
BRND475505
: Brand
Objektglas af klart, optisk glas, hydrolytisk klasse 3.
- Fordybning
- Ant. brønde:1
- Farve:klar
- Beskrivelse:Cavity slides
- Seq:01
- Brønd Ø:15 - 18 mm
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
#: 20864.320
: VWR Chemicals
Acetonitril ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
#: 83640.320
: VWR Chemicals
Natriumhydroxid 0.1 mol/l (0.1 N) in aqueous solution, AVS TITRINORM Reag. Ph. Eur. volumetric solution
#: 31770.294
: VWR Chemicals
Vand, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
#: 83645.320
: VWR Chemicals