Staining troughs with lid
Bakke, med plads til 10 objektglas, 70×91×48 mm
Catalog # BRND472000
Supplier: Brand
Jump to:
Product Details & Documents
Kalk-Soda-Glas
For 10 standard slides, size (76×26 mm)
Ordering information: Please order staining trough, tray, and wire handle seperately.
Specifications
- Beskrivelse:Bakke, med plads til 10 objektglas
- B×D×H:70×91×48 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



