Orders LinkContactUsLinkComponent
Urglas
Urglas
Watch glass, soda-lime glass, with fire-polished edges, 125 mm
Katalog # BRND150065
Leverandør:   Brand
Urglas
Katalog # BRND150065
Leverandør:  Brand
Urglas
Urglas
Watch glass, soda-lime glass, with fire-polished edges, 125 mm
Price
495,00 kr
Log ind for tilgængelighed
1 * 10 Items
Product Details & Documents
Soda-lime glass.
  • Good chemical and physical properties
  • Use in applications that do not involve high thermal stresses
  • Quick and easy to clean
With fire-polished edges. Stress-reduced to minimise risk of shattering.
Certifications: DIN 12341.
Specifikationer
  • Ø:
    125 mm
  • Beskrivelse:
    Watch glass, soda-lime glass, with fire-polished edges