Urglas
Watch glass, soda-lime glass, with fire-polished edges, 80 mm
Katalog # BRND150040
Leverandør: Brand
Gå til:
Product Details & Documents
Soda-lime glass.
- Good chemical and physical properties
- Use in applications that do not involve high thermal stresses
- Quick and easy to clean
With fire-polished edges. Stress-reduced to minimise risk of shattering.
Certifications: DIN 12341.
Specifikationer
- Ø:80 mm
- Beskrivelse:Watch glass, soda-lime glass, with fire-polished edges