Orders LinkContactUsLinkComponent
Urglas
Watch glass, soda-lime glass, with fire-polished edges, 50 mm
  BRND150015
 :   Brand
Urglas
  BRND150015
 :  Brand
Urglas
Urglas
Watch glass, soda-lime glass, with fire-polished edges, 50 mm
272,00 kr
1 * 10 Items
Soda-lime glass.
  • Good chemical and physical properties
  • Use in applications that do not involve high thermal stresses
  • Quick and easy to clean
With fire-polished edges. Stress-reduced to minimise risk of shattering.
 : DIN 12341.
  • Ø:
    50 mm
  • Beskrivelse:
    Watch glass, soda-lime glass, with fire-polished edges