n-Hexan, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Glass bottle, 2,5 L
Leverandør: Avantor
n-Hexan, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Katalog # 8044.2500
Leverandør: Avantor
CAS nummer: 110-54-3
Gå til:
Product Details & Documents
Synonymer: Hexyl hydride, Hexanen, Hexane
- CAS nummer:110-54-3
- Formel:H₃C(CH₂)₄CH₃
- MDL nummer:MFCD00009520
- Molecular Weight:86.18 g/mol
- Merck Index:13,04712
- Kogende Pt:69 °C (1013 hPa)
- Smeltende Pt:–94,3 °C
- Tæthed:0,659 g/cm³ (20 °C)
- Flash Pt:–22 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:203-777-6
- FN:1208
- ADR:3,II
Documents
Certifikater
Søg efter partinummerSpecifikationer
Specifikationstestresultater
- Assay (by GC):min. 95 %
- Aromatic Compounds (as C6H6):max. 0,02 %
- Color (APHA):max. 10
- Density (g/ml) at 25°C:max. 0,657
- Residue after Evaporation:max. 0,001 %
- Sulfur Compounds (as S):max. 0,005 %
- Thiophene:passes test
- Water Soluble Titrable Acid (meq/g):max. 0,0003
- Trace Impurities (in ppm):-
- Aluminium (Al):max. 0,5
- Barium (Ba):max. 0,1
- Boron (B):max. 0,02
- Cadmium (Cd):max. 0,05
- Calcium (Ca):max. 0,5
- Chromium (Cr):max. 0,02
- Cobalt (Co):max. 0,02
- Copper (Cu):max. 0,02
- Iron (Fe):max. 0,1
- Lead (Pb):max. 0,1
- Magnesium (Mg):max. 0,1
- Manganese (Mn):max. 0,02
- Nickel (Ni):max. 0,02
- Tin (Sn):max. 0,1
- Zinc (Zn):max. 0,1
Specifikationer
- Pakning:Glass bottle
- Pakn:2,5 L