Cyclohexan, BAKER ANALYZED® ACS, J.T.Baker®
Glass bottle, 1 L
Leverandør: Avantor
Cyclohexan, BAKER ANALYZED® ACS, J.T.Baker®
Katalog # 8026.1000
Leverandør: Avantor
CAS nummer: 110-82-7
Gå til:
Product Details & Documents
Synonymer: Hexahydrobenzene, Hexamethylene
- CAS nummer:110-82-7
- Formel:C₆H₁₂
- MDL nummer:MFCD00003814
- Molecular Weight:84.16 g/mol
- Merck Index:13,02752
- Kogende Pt:81 °C (1013 hPa)
- Smeltende Pt:7 °C
- Tæthed:0,7785 g/cm³ (20 °C)
- Flash Pt:–18 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:203-806-2
- FN:1145
- ADR:3,II
Specifikationer
Specifikationstestresultater
- Assay (by GC):min. 99,0 %
- Benzene (C6H6):max. 0,005 %
- Color (APHA):max. 10
- Residue after Evaporation:max. 0,002 %
- Substances Darkened by H2SO4:passes test
- Water (H2O):max. 0,02 %
- Trace Impurities (in ppm):-
- Aluminium (Al):max. 0,5
- Barium (Ba):max. 0,1
- Boron (B):max. 0,02
- Cadmium (Cd):max. 0,05
- Calcium (Ca):max. 0,5
- Chromium (Cr):max. 0,02
- Cobalt (Co):max. 0,02
- Copper (Cu):max. 0,02
- Iron (Fe):max. 0,1
- Lead (Pb):max. 0,1
- Magnesium (Mg):max. 0,1
- Manganese (Mn):max. 0,02
- Nickel (Ni):max. 0,02
- Tin (Sn):max. 0,1
- Zinc (Zn):max. 0,1
Specifikationer
- Pakning:Glass bottle
- Pakn:1 L