Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

L (+)-Ascorbinsyre, BAKER ANALYZED® for biotechnology, J.T.Baker®

Plastic bottle for solids, 100 g
Katalog # 1018.0100
Leverandør:   Avantor
L (+)-Ascorbinsyre, BAKER ANALYZED® for biotechnology, J.T.Baker®
Katalog # 1018.0100
Leverandør:  Avantor
CAS nummer:  50-81-7
L (+)-Ascorbinsyre, BAKER ANALYZED® for biotechnology, J.T.Baker®
L (+)-Ascorbinsyre, BAKER ANALYZED® for biotechnology, J.T.Baker®

Plastic bottle for solids, 100 g

CAS nummer:  50-81-7
Price
535,00 kr
Log ind for tilgængelighed
1 * 100 g
Product Details & Documents
Synonymer:  Ascorbic Acid, L-Ascorbic acid, (R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, Acidum ascorbicum
  • CAS nummer:50-81-7
  • Formel:C₆H₈O₆
  • MDL nummer:MFCD00064328
  • Molecular Weight:176.13 g/mol
  • Merck Index:13,00837
  • Kogende Pt:553 °C (1013 hPa)
  • Smeltende Pt:190…192 °C
  • Tæthed:1,694 g/cm³ (20 °C)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:200-066-2
  • NÅ:exempt
Specifikationer

Specifikationstestresultater

  • Assay (dried):
    min. 99,5 %
  • Heavy Metals (as Pb):
    max. 0,001 %
  • Iron (Fe):
    max. 5 ppm
  • Loss on Drying at 105°C:
    max. 0,1 %
  • Residue on Ignition:
    max. 0,05 %
  • Specific Rotation [alpha] 20/D (dried basis), c=10 in water:
    +20.5 - +21.5 °
  • Water Insoluble Material:
    passes test

Specifikationer

  • Pakning:
    Plastic bottle for solids
  • Pakn:
    100 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.