Threaded fittings
Container connection, with outer thread, PP, 19,05 mm
Katalog # BURK8530-0034
Leverandør: BURKLE
Gå til:
Product Details & Documents
Specifikationer
- Gevind:19,05 mm
- Beskrivelse:Container connection, with outer thread
- Materiale:PP
- Seq:29