Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Threaded fittings

Threaded fittings

Sockets, with inner thread, NW 23 mm, PP, 19,05 mm
Katalog # BURK8501-0034
Leverandør:   BURKLE
Threaded fittings
Katalog # BURK8501-0034
Leverandør:  BURKLE
Threaded fittings
Threaded fittings

Sockets, with inner thread, NW 23 mm, PP, 19,05 mm

Price
47,30 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Gevind:
    19,05 mm
  • Beskrivelse:
    Sockets, with inner thread, NW 23 mm
  • Materiale:
    PP
Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.