Accessories for zone samplers
Forseglingsovertræk, PP, steriliserbar, 25 mm Ø, 150 mm, 25 mm
Katalog # 300-0114
Leverandør: BURKLE
Gå til:
Product Details & Documents
Specifikationer
- Ø:25 mm
- Beskrivelse:Forseglingsovertræk, PP, steriliserbar, 25 mm Ø, 150 mm
- Seq:03
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



