Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Natriumhydroxid, BAKER, J.T.Baker®

Natriumhydroxid, BAKER, J.T.Baker®

Plastic bottle for solids, 5 kg
Katalog # 0405.5000
Ætsende
Leverandør:   Avantor
Natriumhydroxid, BAKER, J.T.Baker®
Katalog # 0405.5000
Leverandør:  Avantor
CAS nummer:  1310-73-2
Natriumhydroxid, BAKER, J.T.Baker®

Plastic bottle for solids, 5 kg

CAS nummer:  1310-73-2
Price
1.240,00 kr
Log ind for tilgængelighed
1 * 5 kg
Product Details & Documents
Synonymer:  Caustic soda
  • CAS nummer:1310-73-2
  • Formel:NaOH
  • MDL nummer:MFCD00003548
  • Molecular Weight:40 g/mol
  • Merck Index:14,08627
  • Kogende Pt:1390 °C (1013 hPa)
  • Smeltende Pt:323 °C
  • Tæthed:2,13 g/cm³ (20 °C)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:215-185-5
  • FN:1823
  • ADR:8,II
  • NÅ:01-2119457892-27
Specifikationer

Specifikationstestresultater

  • Assay:
    97.0-100.5 %
  • Appearance of solution:
    passes test
  • Carbonate (as Na2CO3):
    max. 2,0 %
  • Chlorides (as Cl):
    max. 50 ppm
  • Heavy Metals (as Pb):
    max. 20 ppm
  • Identification:
    passes test
  • Iron (Fe):
    max. 10 ppm
  • Sulfates (as SO4):
    max. 50 ppm
  • Additional Specification(s)::
    -
  • Potassium (K):
    passes test

Specifikationer

  • Pakning:
    Plastic bottle for solids
  • Pakn:
    5 kg

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.