Digler
5 ml, 20×25 mm, 0,5 mm
BOCH8100
: BOCHEM
These crucibles and lids are made of nickel or stainless steel 18/10.
: Lids need to be ordered separately.
- Diameter×H:20×25 mm
- Kapacitet:5 ml
- Seq:01
- Tykkelse:0,5 mm
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
#: 20864.320
: VWR Chemicals
Acetonitril ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
#: 83640.320
: VWR Chemicals
Natriumhydroxid 0.1 mol/l (0.1 N) in aqueous solution, AVS TITRINORM Reag. Ph. Eur. volumetric solution
#: 31770.294
: VWR Chemicals
Vand, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
#: 83645.320
: VWR Chemicals