Stand bases
2,0 kg, 160×250×6 mm
Katalog # BOCH5032
Leverandør: BOCHEM
Gå til:
Product Details & Documents
Steel, varnished.
- With M10 threaded hole
- One adjustable foot
Certifications: DIN 12892
Specifikationer
- B×D×H:160×250×6 mm
- Vægt:2,0 kg
Steel, varnished.
Certifications: DIN 12892