pH probes
pH probe, glass, for testo 206 pH3, with temperature sensor, 0 - 14, 0...80 °C, Gel, 12×120 mm, General purposes
Katalog # 663-3215
Leverandør: Testo
Gå til:
Product Details & Documents
Specifikationer
- Ø×L:12×120 mm
- Til:General purposes
- Beskrivelse:pH probe, glass, for testo 206 pH3, with temperature sensor
- Seq:01
- Elektrolyt:Gel
- Temperatur:0...80 °C
- pH:0 - 14