Burettes, lateral stopcock, class AS, Schellbach
25 ml, 0,05 ml, 0,03 ± ml
Katalog # 661-4121
Leverandør: witeg Labortechnik
Gå til:
Product Details & Documents
Burettes, class AS, Schellbach blue line, conformity certified and made of clear borosilicate glass.
- Main point ring graduation
- Lateral stopcock, with PTFE-needle valve
- Overtwist device
Specifikationer
- Kapacitet:25 ml
- Inddeling:0,05 ml
- Tolerance:0,03 ± ml