Orders LinkContactUsLinkComponent
Formalingskugler
Formalingskugler
Formalingskugler, Sintered aluminium oxide, 30 mm
Catalog # 412-2882
Supplier:   Retsch GmbH
Formalingskugler
Catalog # 412-2882
Supplier:  Retsch GmbH
Formalingskugler
Formalingskugler
Formalingskugler, Sintered aluminium oxide, 30 mm
Price
706,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Specifications
  • Ø:
    30 mm
  • Beskrivelse:
    Formalingskugler
  • Materiale:
    Sintered aluminium oxide
  • Seq:
    17
Accessories
Product Information
  • Catalog #
    Beskrivelse
    Materiale
    Ø
    Availability
    Price
  • 412-2912
    Grinding ball
    Agat
    5 mm
    Log in for availability
    1 * 1 Items - 66,40 kr
Product Information
  • Catalog #
    Beskrivelse
    Materiale
    Ø
    Availability
    Price
  • 412-3131
    Grinding ball
    Hærdet stål
    5 mm
    Log in for availability
    1 * 1 Items - 15,60 kr
Product Information
  • Catalog #
    Beskrivelse
    Materiale
    Ø
    Availability
    Price
  • 412-3141
    Grinding ball
    Rustfrit stål
    5 mm
    Log in for availability
    1 * 1 Items - 16,60 kr
Product Information
  • Catalog #
    Beskrivelse
    Materiale
    Ø
    Availability
    Price
  • 412-3151
    Grinding ball
    Wolframcarbid
    5 mm
    Log in for availability
    1 * 1 Items - 177,00 kr