Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Kaliumchlorid, EMSURE® for analysis, Supelco®

10 kg
  1.04936.9010
 :   Merck
Kaliumchlorid, EMSURE® for analysis, Supelco®
  1.04936.9010
 :  Merck
 :  7447-40-7
Kaliumchlorid, EMSURE® for analysis, Supelco®

10 kg

 :  7447-40-7
 
4.850,00 kr
   
1 * 10 kg
 
 :  Potassiumchloride
  •  :7447-40-7
  •  :KCl
  •  :MFCD00011360
  •  :74.55 g/mol
  •  :14,07621
  •  :1420 °C (1013 hPa)
  •  :773 °C
  •  :1,98 g/cm³ (20 °C)
  •  :M-Clarity™ quality level = MQ300
  •  :Stue temperatur
  •  :231-211-8
 

 

  • Solubility:
    347 g/l (20 °C)
  • Melting Point:
    773 °C
  • Molar Mass:
    74.55 g/mol
  • Bulk Density:
    1000 kg/m3
  • Boiling Point:
    1413 °C (1013 hPa)
  • Density:
    1.98 g/cm3 (20 °C)
  • pH:
    5.5 - 8.0 (50 g/l, H2O, 25 °C)

 

  • Pakn:
    10 kg