Probes for 445
Pitot tube, stainless steel, for measuring flow velocity, 1...+100 m/s, —, 7×300 mm
Katalog # 628-3141
Leverandør: Testo
Gå til:
Product Details & Documents
Specifikationer
- Område:1...+100 m/s
- Beskrivelse:Pitot tube, stainless steel, for measuring flow velocity
- Seq:06
- Nøjagtighed:—
- Ø×L:7×300 mm