Probes for 445
Pitot tube, stainless steel, for measuring flow velocity, 1...+100 m/s, —, 7×1000 mm
Catalog # 628-3139
Supplier: Testo
Jump to:
Product Details & Documents
Specifications
- Område:1...+100 m/s
- Beskrivelse:Pitot tube, stainless steel, for measuring flow velocity
- Seq:08
- Nøjagtighed:—
- Ø×L:7×1000 mm