Phenol rød ACS indicator, Fluka™
Glass bottle, 100 g
Katalog # 32661-100G
Leverandør: Honeywell Chemicals
Phenol rød ACS indicator, Fluka™
Katalog # 32661-100G
Leverandør: Honeywell Chemicals
CAS nummer: 143-74-8
Gå til:
Product Details & Documents
Synonymer: 3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
- CAS nummer:143-74-8
- Formel:C₁₉H₁₄O₅S
- MDL nummer:MFCD00003552
- Molecular Weight:354.38 g/mol
- Merck Index:13,07329
- Smeltende Pt:> 300 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:205-609-7
Documents
Certifikater
Søg efter partinummerSpecifikationer
Specifikationstestresultater
- Clearness of the solution:complying
- Loss on drying (vacuum):Max. 5 %
- Residue on ignition:Max. 1 %
- Change range:complying
- Extinction maximum at:552 - 562 nm
- Extinction maximum of:900 - 1800
Specifikationer
- Pakning:Glass bottle
- Pakn:100 g
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



