Phenol rød ACS indicator, Fluka™
Glass bottle, 10 g
Catalog # 32661-10G
Supplier: Honeywell Chemicals
Phenol rød ACS indicator, Fluka™
Catalog # 32661-10G
Supplier: Honeywell Chemicals
CAS Number: 143-74-8
Jump to:
Product Details & Documents
Synonyms: 3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
- CAS Number:143-74-8
- Formula:C₁₉H₁₄O₅S
- MDL Number:MFCD00003552
- Molecular Weight:354.38 g/mol
- Merck Index:13,07329
- Melting Pt:> 300 °C
- Storage Temperature:Stue temperatur
- EINECS:205-609-7
Documents
Certificates
Search by Lot NumberSpecifications
Specification Test Results
- Clearness of the solution:complying
- Loss on drying (vacuum):Max. 5 %
- Residue on ignition:Max. 1 %
- Change range:complying
- Extinction maximum at:552 - 562 nm
- Extinction maximum of:900 - 1800
Specifications
- Pakning:Glass bottle
- Pakn:10 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



