Pipette tips, ART™
ART™ 10, 10 µl, -, 31,3 mm, Pose
Katalog # 612-9751
Leverandør: Thermo Fisher Scientific
Pipette tips, ART™
Katalog # 612-9751
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
These non filtered tips are designed to fit virtually any micropipette. They are suitable for use when working with the most difficult samples, such as radioactive compounds, viscous materials, fragile cell lines, viruses, difficult proteins and PCR reagents.
- Non sterile tips are certified to be free of RNase, DNase, DNA, ATP, endotoxins, pyrogens and PCR inhibitors
- Sterile tips are certified to be free of RNase, DNase, DNA, ATP, bioburden, endotoxins, pyrogens and PCR inhibitors
- Rack cover can easily be realigned from a traditional lift-off to a hinged rack for one-handed access to tips
- Front latch closes and locks ensure safe and secure storage
- Space saving reload system is environmentally friendly, traceable and easy to use
Emballering: Tips are available in bulk packaging, in racks or in trays for rack reloading. Reload tips require an empty rack.
Documents
Specifikationer
- Pakket:Pose
- Steril:-
- Volumen:10 µl
- Længde:31,3 mm
- Seq:01
- Spidstype:ART™ 10
- Cat. No.:612-9751
- Miljøcertificering:IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- Environmentally Preferable:Lav fremstillingspåvirkning
Accessories
Produktinformation
- Katalog #TilgængelighedPrice
- Tomt rack til refillpipettespidser, MediumTomt rack til refillpipettespidser, Medium732-1170Log ind for tilgængelighed1 * 10 Items - 273,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- Tomt rack til refillpipettespidser, LargeTomt rack til refillpipettespidser, Large732-1169Log ind for tilgængelighed1 * 8 Items - 134,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- Tomt rack til refillpipettespidser, SmallTomt rack til refillpipettespidser, Small732-1171Log ind for tilgængelighed1 * 10 Items - 153,00 kr
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



