Orders LinkContactUsLinkComponent
Inoculating loops and needles, disposable
Platinøse, 10 µl, blå, pakket enkeltvis
  391-2082
Inoculating loops and needles, disposable
  391-2082
Inoculating loops and needles, disposable
Inoculating loops and needles, disposable
Platinøse, 10 µl, blå, pakket enkeltvis
1.260,00 kr
1 * 600 Items
Flexible polymer.
  • Sterilised by irradiation
  • Individually wrapped
  • Length: 200 mm
Documents
  • Pakket:
    pakket enkeltvis
  • Kapacitet:
    10 µl
  • Farve:
    blå
  • Beskrivelse:
    Platinøse
  • Seq:
    04