Pipette tips, ratiolab®
Finntip, 1000 - 5000 µl, -, Transparent, Finnpipette F1, F2 og Dr. Lange-pipetter, 75 stk./sammenklappelig æske
612-1484
: RATIOLAB
ratiolab® pipette tips are ideal for use with all commercially available pipettes.
- Made of high quality PP
- Tight fit due to the precisely fitting cone
- Guaranteed dust-free due to special ionization treatment
- Excellent transparency due to high quality surfaces
- Minimal residual wetting due to controlled quality of raw materials
- Excellent volume accuracy due to precise tip orifice
- MultiTray® refill system is space- and cost-saving and clearly reduces the amount of waste
MultiTray®: Dust-proof refill pack for the Multibox® plus and the Multibox® ultra.
- Pakket:75 stk./sammenklappelig æske
- Steril:-
- Volumen:1000 - 5000 µl
- Seq:44
- Spidstype:Finntip
- Spits farge:Transparent
- Til brug med:Finnpipette F1, F2 og Dr. Lange-pipetter