Orders LinkContactUsLinkComponent
Pipette tips, ratiolab®
Pipette tips, ratiolab®
Finntip, 1000 - 5000 µl, -, Transparent, Finnpipette F1, F2 og Dr. Lange-pipetter, 75 stk./sammenklappelig æske
  612-1484
 :   RATIOLAB
Pipette tips, ratiolab®
  612-1484
 :  RATIOLAB
Pipette tips, ratiolab®
Pipette tips, ratiolab®
Finntip, 1000 - 5000 µl, -, Transparent, Finnpipette F1, F2 og Dr. Lange-pipetter, 75 stk./sammenklappelig æske
170,00 kr
1 * 75 Items
ratiolab® pipette tips are ideal for use with all commercially available pipettes.
  • Made of high quality PP
  • Tight fit due to the precisely fitting cone
  • Guaranteed dust-free due to special ionization treatment
  • Excellent transparency due to high quality surfaces
  • Minimal residual wetting due to controlled quality of raw materials
  • Excellent volume accuracy due to precise tip orifice
  • MultiTray® refill system is space- and cost-saving and clearly reduces the amount of waste
MultiTray®: Dust-proof refill pack for the Multibox® plus and the Multibox® ultra.
  • Pakket:
    75 stk./sammenklappelig æske
  • Steril:
    -
  • Volumen:
    1000 - 5000 µl
  • Seq:
    44
  • Spidstype:
    Finntip
  • Spits farge:
    Transparent
  • Til brug med:
    Finnpipette F1, F2 og Dr. Lange-pipetter