Pipette tips, ratiolab®
Type E, 0,5 - 20 µl, -, Transparent, almindeligt anvendte pipetter, 1.000 stk./pose
Katalog # 612-1477
Leverandør: RATIOLAB
Gå til:
Product Details & Documents
ratiolab® pipette tips are ideal for use with all commercially available pipettes.
- Made of high quality PP
- Tight fit due to the precisely fitting cone
- Guaranteed dust-free due to special ionization treatment
- Excellent transparency due to high quality surfaces
- Minimal residual wetting due to controlled quality of raw materials
- Excellent volume accuracy due to precise tip orifice
- MultiTray® refill system is space- and cost-saving and clearly reduces the amount of waste
MultiTray®: Dust-proof refill pack for the Multibox® plus and the Multibox® ultra.
Specifikationer
- Pakket:1.000 stk./pose
- Steril:-
- Volumen:0,5 - 20 µl
- Seq:14
- Spidstype:Type E
- Spits farge:Transparent
- Til brug med:almindeligt anvendte pipetter