Orders LinkContactUsLinkComponent
Volumetric flasks, class A, white markings
Volumetric flasks, class A, white markings
200 ml, 14/23
Catalog # 612-0563
Supplier:   Poulten & Graf
Volumetric flasks, class A, white markings
Catalog # 612-0563
Supplier:  Poulten & Graf
Volumetric flasks, class A, white markings
Volumetric flasks, class A, white markings
200 ml, 14/23
Price
388,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Borosilicate glass, amber, with PP stopper, conformity certified.
  • With ground ring mark
  • White markings
Specifications
  • NS:
    14/23
  • Kapacitet:
    200 ml