Volumetric flasks, class A, white markings
200 ml, 14/23
Catalog # 612-0563
Supplier: Poulten & Graf
Jump to:
Product Details & Documents
Borosilicate glass, amber, with PP stopper, conformity certified.
- With ground ring mark
- White markings
Specifications
- NS:14/23
- Kapacitet:200 ml