Screw caps with liners, WHEATON®
Skruelåg, GPI-gevindafslutning 20 - 400, White PP, solid top, Foamed PE
Katalog # 548-2041
Leverandør: DWK Life Sciences
Gå til:
Product Details & Documents
These PP screw caps have multi-purpose liners, which are made of three-ply co-extruded foamed low density PE core between two solid layers of low density PE.
- For use with neutral pH to mild acid and alkali solutions
Specifikationer
- Septum:Foamed PE
- Model:White PP, solid top
- Beskrivelse:Skruelåg, GPI-gevindafslutning 20 - 400
- Seq:02