Slanger
0,50 mm, 1/16"=1,6 mm
Katalog # 554-1096
Leverandør: VICI JOUR RESEARCH
Gå til:
Product Details & Documents
SS316 svejset under inert gas.
- Smooth internal surface
- Soft annealed OD for easy ferrule connections
En bred vifte af størrelser til HPLC og GC applikationer. Slanger kan modstå en trykeffekt på min. 315 bar (4600 psi).
Specifikationer
- Udv.Ø:1/16"=1,6 mm
- Ø indv.:0,50 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



