Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Incinerating dishes, rectagular

50 mm, 20 mm, 20 mm
Katalog # 539-1924
Incinerating dishes, rectagular
Katalog # 539-1924
Incinerating dishes, rectagular
Incinerating dishes, rectagular

50 mm, 20 mm, 20 mm

Price
448,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

Alsint 99,7.

  • Good thermal shock resistance due to high thermal conductivity
  • Resistant to chemical attack, e.g. hydrogen and other reducing gases
Specifikationer
  • Bredde:
    20 mm
  • Højde:
    20 mm
  • Længde:
    50 mm
  • Seq:
    02