Test tube baskets
152×152×152 mm
Katalog # 489-2211
Leverandør: Bel-Art Products, a Part of SP
Gå til:
Product Details & Documents
PP.
- Very good chemical resistance
- Very good temperature resistance
- Sterilisable and autoclavable
These test tube baskets are designed for rinsing, transporting and storing laboratory glassware.
Colour: White
Specifikationer
- B×D×H:152×152×152 mm