Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Copper phosphorus shot

Copper phosphorus shot

2 kg
Katalog # 88357.A3
Leverandør:   Thermo Fisher Scientific
Copper phosphorus shot
Katalog # 88357.A3
CAS nummer:  
Copper phosphorus shot

2 kg

CAS nummer:  
Price
6.160,00 kr
Log ind for tilgængelighed
1 * 2 kg
Product Details & Documents

Molecular Formula: Cu:P; 85:15 wt% MDL No.: MFCD01074956

  • Formel:Cu:P;₈₅:₁₅%
  • MDL nummer:MFCD01074956
  • Opbevaringstemperatur:Stue temperatur

Documents

Specifikationer

Specifikationer

  • Pakn:
    2 kg

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.